2-Nitro-5-piperidinobenzenecarbohydrazide ≥95%

Supplier: Matrix Scientific

046120-500MG
101874-256EA 556.92 USD
101874-256
2-Nitro-5-piperidinobenzenecarbohydrazide ≥95%
2-Nitro-5-piperidinobenzenecarbohydrazide

Hazard: Irritant

Formula: C₁₂H₁₆N₄O₃
Melting Pt: 155…157 °C
MDL Number: MFCD12025870
CAS Number: 1147979-35-8

Order Now

Specification Test Results

Melting point 155-157°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR