6-(Trifluoromethyl)imidazo[1,2-a]pyridine ≥95%
Supplier: Matrix Scientific
046105-500MG
046105-1G
046105-5G
101874-188EA
160
USD
101874-188
101874-192
101874-190
6-(Trifluoromethyl)imidazo[1,2-a]pyridine ≥95%
6-(Trifluoromethyl)imidazo[1,2-a]pyridine
Hazard: Irritant
Formula:
C₈H₅F₃N₂ Melting Pt: 98…99 °C |
MDL Number:
MFCD09994631 CAS Number: 936009-02-8 |
Specification Test Results
Melting point | 98-99°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...