2-Chlorophenacyl thiocyanate ≥95%
Supplier: Matrix Scientific
046092-500MG
046092-5G
046092-1G
101874-130EA
806.15
USD
101874-130
101874-128
101874-126
2-Chlorophenacyl thiocyanate ≥95%
2-Chlorophenacyl thiocyanate
Hazard: Irritant
Formula:
C₉H₆ClNOS Melting Pt: 56…58 °C |
MDL Number:
MFCD09152712 CAS Number: 19339-58-3 |
Specification Test Results
Melting point | 56-58°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...