5-Nitro-2-(2-propynylsulfanyl)pyridine ≥95%
Supplier: Matrix Scientific
046088-1G
046088-500MG
101874-110EA
349.6
USD
101874-110
101874-108
5-Nitro-2-(2-propynylsulfanyl)pyridine ≥95%
5-Nitro-2-(2-propynylsulfanyl)pyridine
Hazard: Irritant
Formula:
C₈H₆N₂O₂S MW: 194.21 g/mol Melting Pt: 102…104 °C |
MDL Number:
MFCD09152764 CAS Number: 77867-13-1 |
Specification Test Results
Melting point | 102-104°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...