7-Acetyl-2-ethylbenzoxazole ≥95%

Supplier: Matrix Scientific

Synonyms: 1-(2-Ethyl-1,3-benzoxazol-7-yl)ethan-1-one

046085-5G 046085-500MG 046085-1G
101874-094EA 806.15 USD
101874-094 101874-092 101874-090
7-Acetyl-2-ethylbenzoxazole ≥95%
7-Acetyl-2-ethylbenzoxazole

Hazard: Irritant

Formula: C₁₁H₁₁NO₂
Melting Pt: 42…44 °C
MDL Number: MFCD09152709
CAS Number: 952182-97-7

Order Now

Specification Test Results

Melting point 42-44°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR