4-Cyclopropylisoxazolo[3,4-d]pyridazin-7(6H)-one ≥95%
Supplier: Matrix Scientific
046082-500MG
046082-1G
101874-078EA
707.69
USD
101874-078
101874-076
4-Cyclopropylisoxazolo[3,4-d]pyridazin-7(6H)-one ≥95%
4-Cyclopropylisoxazolo[3,4-d]pyridazin-7(6H)-one
Hazard: Irritant
Formula:
C₈H₇N₃O₂ Melting Pt: 210…212 °C |
MDL Number:
MFCD09152706 CAS Number: 952182-94-4 |
Specification Test Results
Melting point | 210-212°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...