4-(3,4-Dihydro-2H-1,5-benzodioxepin-7-yl)benzenecarbaldehyde ≥95%
Supplier: Matrix Scientific
Synonyms:
4-(3,4-Dihydro-2H-1,5-benzodioxepin-7-yl)benzaldehyde
046079-1G
046079-500MG
101874-064EA
467.69
USD
101874-064
101874-062
4-(3,4-Dihydro-2H-1,5-benzodioxepin-7-yl)benzenecarbaldehyde ≥95%
4-(3,4-Dihydro-2H-1,5-benzodioxepin-7-yl)benzenecarbaldehyde
Hazard: Irritant
Formula:
C₁₆H₁₄O₃ MW: 254.29 g/mol Melting Pt: 94…96 °C |
MDL Number:
MFCD09152704 CAS Number: 952182-92-2 |
Specification Test Results
Melting point | 94-96°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...