7-Acetylbenzoxazole ≥95%
Supplier: Matrix Scientific
Synonyms:
1-(1,3-Benzoxazol-7-yl)ethan-1-one
046075-500MG
046075-1G
046075-5G
101874-044EA
564.62
USD
101874-044
101874-042
101874-040
7-Acetylbenzoxazole ≥95%
7-Acetylbenzoxazole
Hazard: Irritant
Formula:
C₉H₇NO₂ Melting Pt: 118…120 °C |
MDL Number:
MFCD09152701 CAS Number: 952182-89-7 |
Specification Test Results
Melting point | 118-120°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...