Ethyl-3-formyl-1H-indole-2-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 3-formyl-1H-indole-2-carboxylate

046064-1G 046064-500MG 046064-5G
101873-982EA 923.08 USD
101873-982 101873-978 101873-980
Ethyl-3-formyl-1H-indole-2-carboxylate ≥95%
Ethyl-3-formyl-1H-indole-2-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₁NO₃
MW: 217.23 g/mol
Melting Pt: 186…188 °C
MDL Number: MFCD01463252
CAS Number: 18450-27-6

Order Now

Specification Test Results

Melting point 186-188°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR