2-Bromo-4-methoxy-6-nitrobenzenol ≥95%
Supplier: Matrix Scientific
Synonyms:
2-Bromo-4-methoxy-6-nitrophenol
046051-1G
046051-500MG
046051-5G
101873-920EA
232.31
USD
101873-920
101873-922
101873-918
2-Bromo-4-methoxy-6-nitrobenzenol ≥95%
2-Bromo-4-methoxy-6-nitrobenzenol
Hazard: Irritant
Formula:
C₇H₆BrNO₄ MW: 248.04 g/mol Melting Pt: 116…118 °C |
MDL Number:
MFCD06797984 CAS Number: 115929-59-4 |
Specification Test Results
Melting point | 116-118°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...