3-Amino-4-methylbenzenecarbohydrazide ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Amino-4-methylbenzhydrazide

046033-500MG 046033-5G 046033-1G
101873-830EA 232.31 USD
101873-830 101873-832 101873-828
3-Amino-4-methylbenzenecarbohydrazide ≥95%
3-Amino-4-methylbenzenecarbohydrazide

Hazard: Irritant

Formula: C₈H₁₁N₃O
MW: 165.2 g/mol
Melting Pt: 128…130 °C
MDL Number: MFCD08276933
CAS Number: 845884-82-4

Order Now

Specification Test Results

Melting point 128-130°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR