7-Bromo-5-methoxy-1,3-benzoxazole ≥95%

Supplier: Matrix Scientific

046027-1G
101873-808EA 698.46 USD
101873-808
7-Bromo-5-methoxy-1,3-benzoxazole ≥95%
7-Bromo-5-methoxy-1,3-benzoxazole

Hazard: Irritant

Formula: C₈H₆BrNO₂
Melting Pt: 107…109 °C
MDL Number: MFCD08689669
CAS Number: 937601-57-5

Order Now

Specification Test Results

Melting point 107-109°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR