2H-Pyrido[3,2-b][1,4]oxazin-3(4H)-one ≥95%

Supplier: Matrix Scientific

046014-1G 046014-5G 046014-500MG
101873-742EA 135.7 USD
101873-742 101873-744 101873-746
2H-Pyrido[3,2-b][1,4]oxazin-3(4H)-one ≥95%
2H-Pyrido[3,2-b][1,4]oxazin-3(4H)-one

Hazard: Irritant

Formula: C₇H₆N₂O₂
Melting Pt: 204…207 °C
MDL Number: MFCD00006697
CAS Number: 20348-09-8

Order Now

Specification Test Results

Melting point 204-207°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR