2-(4-Fluorophenyl)nicotinaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(4-Fluorophenyl)pyridine-3-carbaldehyde

046010-500MG 046010-1G 046010-5G
101873-720EA 601.54 USD
101873-720 101873-722 101873-724
2-(4-Fluorophenyl)nicotinaldehyde ≥95%
2-(4-Fluorophenyl)nicotinaldehyde

Hazard: Irritant

Formula: C₁₂H₈FNO
MW: 201.2 g/mol
Melting Pt: 88…90 °C
MDL Number: MFCD06659613
CAS Number: 885950-17-4

Order Now

Specification Test Results

Melting point 88-90°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR