6-(4-Formylphenyl)nicotinaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 6-(4-Formylphenyl)pyridine-3-carbaldehyde

046003-500MG 046003-1G 046003-5G
101873-680EA 747.69 USD
101873-680 101873-682 101873-684
6-(4-Formylphenyl)nicotinaldehyde ≥95%
6-(4-Formylphenyl)nicotinaldehyde

Hazard: Irritant

Formula: C₁₃H₉NO₂
MW: 211.22 g/mol
Melting Pt: 155…158 °C
MDL Number: MFCD06659608
CAS Number: 885950-11-8

Order Now

Specification Test Results

Melting point 155-158°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR