Ethyl-8-fluoro-1,3,4,5-tetrahydro-2H-pyrido[4,3-b]indole-2-carboxylate ≥95%
Supplier: Matrix Scientific
046000-5G
046000-1G
046000-500MG
101873-664EA
230.77
USD
101873-664
101873-666
101873-662
Ethyl-8-fluoro-1,3,4,5-tetrahydro-2H-pyrido[4,3-b]indole-2-carboxylate ≥95%
Ethyl-8-fluoro-1,3,4,5-tetrahydro-2H-pyrido[4,3-b]indole-2-carboxylate
Hazard: Irritant
Formula:
C₁₄H₁₅FN₂O₂ MW: 262.29 g/mol Melting Pt: 172…173 °C |
MDL Number:
MFCD02930345 CAS Number: 58038-66-7 |
Specification Test Results
Melting point | 172-173°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...