2-Bromo-4'-(methylsulfonyl)acetophenone ≥91%

Supplier: Matrix Scientific

Synonyms: 2-Bromo-1-(4-(methylsulphonyl)phenyl)ethanone4-(Methylsulphonyl)phenacyl bromide2-Bromo-1-(4-(methylsulfonyl)phenyl)ethanone4-(Methylsulfonyl)phenacyl bromide

045984-5G
101873-594EA 52.31 USD
101873-594
2-Bromo-4'-(methylsulfonyl)acetophenone ≥91%
2-Bromo-4'-(methylsulphonyl)acetophenone

Hazard: Irritant

Formula: C₉H₉BrO₃S
MW: 277.1350 g/mol
Melting Pt: 126…128 °C
MDL Number: MFCD00673134
CAS Number: 50413-24-6

Order Now

Specification Test Results

Melting point 126-128°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR