3-Chloro-2,3-bis(4-methoxyphenyl)acrylaldehyde ≥95%

Supplier: Matrix Scientific

045974-5G 045974-1G 045974-500MG
101873-542EA 200 USD
101873-542 101873-544 101873-546
3-Chloro-2,3-bis(4-methoxyphenyl)acrylaldehyde ≥95%
3-Chloro-2,3-bis(4-methoxyphenyl)acrylaldehyde

Hazard: Irritant

Formula: C₁₇H₁₅ClO₃
Melting Pt: 160…163 °C
MDL Number: MFCD00547134
CAS Number: 19881-70-0

Order Now

Specification Test Results

Melting point 160-163°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR