3-(4-Hydroxybenzyl)-2,4-pentanedione dioxime ≥95%

Supplier: Matrix Scientific

045973-500MG 045973-1G 045973-5G
101873-536EA 278.46 USD
101873-536 101873-538 101873-540
3-(4-Hydroxybenzyl)-2,4-pentanedione dioxime ≥95%
3-(4-Hydroxybenzyl)-2,4-pentanedione dioxime

Hazard: Irritant

Formula: C₁₂H₁₆N₂O₃
Melting Pt: 210…212 °C
MDL Number: MFCD03425829
CAS Number: 454473-86-0

Order Now

Specification Test Results

Melting point 210-212°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR