Methyl N-(4-methoxyphenyl)carbamate ≥95%

Supplier: Matrix Scientific

045971-500MG 045971-5G 045971-1G
101873-524EA 278.46 USD
101873-524 101873-526 101873-528
Methyl N-(4-methoxyphenyl)carbamate ≥95%
Methyl N-(4-methoxyphenyl)carbamate

Hazard: Irritant

Formula: C₉H₁₁NO₃
Melting Pt: 88…89 °C
MDL Number: MFCD00025852
CAS Number: 14803-72-6

Order Now

Specification Test Results

Melting point 88-89°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR