2-[3-(4-Nitrophenyl)acryloyl]benzenecarboxylic acid ≥95%

Supplier: Matrix Scientific

045964-5G 045964-500MG 045964-1G
101873-490EA 135.7 USD
101873-490 101873-492 101873-494
2-[3-(4-Nitrophenyl)acryloyl]benzenecarboxylic acid ≥95%
2-[3-(4-Nitrophenyl)acryloyl]benzenecarboxylic acid

Hazard: Irritant

Formula: C₁₆H₁₁NO₅
Melting Pt: 187…189 °C
MDL Number: MFCD03617865
CAS Number: 6261-66-1

Order Now

Specification Test Results

Melting point 187-189°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR