5-Fluoroisophthalonitrile ≥95%

Supplier: Matrix Scientific

045958-5G
101873-460EA 136.92 USD
101873-460
5-Fluoroisophthalonitrile ≥95%
5-Fluoroisophthalonitrile

Hazard: Irritant

Formula: C₈H₃FN₂
Melting Pt: 109…111 °C
MDL Number: MFCD05663831
CAS Number: 453565-55-4

Order Now

Specification Test Results

Melting point 109-111°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR