3-Fluoro-2-methyl-6-nitroaniline ≥95%

Supplier: Matrix Scientific

045952-1G 045952-5G 045952-500MG
101873-436EA 350.77 USD
101873-436 101873-438 101873-440
3-Fluoro-2-methyl-6-nitroaniline ≥95%
3-Fluoro-2-methyl-6-nitroaniline

Hazard: Irritant

Formula: C₇H₇FN₂O₂
Melting Pt: 118…121 °C
MDL Number: MFCD03787364
CAS Number: 485832-96-0

Order Now

Specification Test Results

Melting point 118-121°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR