3-[(tert-Butoxycarbonyl)amino]-3-(4-hydroxyphenyl)propanoic acid ≥95%
Supplier: Matrix Scientific
045947-1G
045947-5G
045947-500MG
101873-410EA
692.31
USD
101873-410
101873-412
101873-408
3-[(tert-Butoxycarbonyl)amino]-3-(4-hydroxyphenyl)propanoic acid ≥95%
3-[(tert-Butoxycarbonyl)amino]-3-(4-hydroxyphenyl)propanoic acid
Hazard: Irritant
Formula:
C₁₄H₁₉NO₅ Melting Pt: 143…145 °C |
MDL Number:
MFCD03001312 CAS Number: 454473-84-8 |
Specification Test Results
Melting point | 143-145°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...