3-[(tert-Butoxycarbonyl)amino]-3-[4-(methylsulfanyl)phenyl]propanoic acid ≥95%

Supplier: Matrix Scientific

045946-500MG 045946-1G
101873-404EA 556.92 USD
101873-404 101873-406
3-[(tert-Butoxycarbonyl)amino]-3-[4-(methylsulfanyl)phenyl]propanoic acid ≥95%
3-[(tert-Butoxycarbonyl)amino]-3-[4-(methylsulfanyl)phenyl]propanoic acid

Hazard: Irritant

Formula: C₁₅H₂₁NO₄S
Melting Pt: 166…168 °C
MDL Number: MFCD03001311
CAS Number: 454473-83-7

Order Now

Specification Test Results

Melting point 166-168°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR