3-(1-Oxo-1,3-dihydro-2H-isoindol-2-yl)-3-(3-thienyl)propanoic acid ≥95%

Supplier: Matrix Scientific

045944-500MG 045944-1G 045944-5G
101873-392EA 747.69 USD
101873-392 101873-394 101873-396
3-(1-Oxo-1,3-dihydro-2H-isoindol-2-yl)-3-(3-thienyl)propanoic acid ≥95%
3-(1-Oxo-1,3-dihydro-2H-isoindol-2-yl)-3-(3-thienyl)propanoic acid

Hazard: Irritant

Formula: C₁₅H₁₃NO₃S
Melting Pt: 166…168 °C
MDL Number: MFCD02571531
CAS Number: 478249-84-2

Order Now

Specification Test Results

Melting point 166-168°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR