3-(4-Methylphenyl)-3-(1-oxo-1,3-dihydro-2H-isoindol-2-yl)propanoic acid ≥95%
Supplier: Matrix Scientific
045943-500MG
045943-5G
045943-1G
101873-388EA
467.69
USD
101873-388
101873-390
101873-386
3-(4-Methylphenyl)-3-(1-oxo-1,3-dihydro-2H-isoindol-2-yl)propanoic acid ≥95%
3-(4-Methylphenyl)-3-(1-oxo-1,3-dihydro-2H-isoindol-2-yl)propanoic acid
Hazard: Irritant
Formula:
C₁₈H₁₇NO₃ MW: 295.34 g/mol Melting Pt: 181…183 °C |
MDL Number:
MFCD03425823 CAS Number: 439095-70-2 |
Specification Test Results
Melting point | 181-183°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...