3-(5-Bromo-2-thienyl)-3-[(2,2,2-trifluoroacetyl)amino]propanoic acid ≥95%
Supplier: Matrix Scientific
045942-1G
045942-5G
045942-500MG
101873-380EA
375.38
USD
101873-380
101873-382
101873-384
3-(5-Bromo-2-thienyl)-3-[(2,2,2-trifluoroacetyl)amino]propanoic acid ≥95%
3-(5-Bromo-2-thienyl)-3-[(2,2,2-trifluoroacetyl)amino]propanoic acid
Hazard: Irritant
Formula:
C₉H₇BrF₃NO₃S Melting Pt: 168…170 °C |
MDL Number:
MFCD03425822 CAS Number: 194229-21-5 |
Specification Test Results
Melting point | 168-170°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...