2-(4-Morpholino)-5-nitropyridine ≥95%
Supplier: Matrix Scientific
Synonyms:
4-(5-Nitropyridin-2-yl)morpholine
045933-500MG
045933-5G
045933-1G
101873-336EA
18.46
USD
101873-336
101873-338
101873-340
2-(4-Morpholino)-5-nitropyridine ≥95%
2-(4-Morpholino)-5-nitropyridine
Hazard: Irritant
Formula:
C₉H₁₁N₃O₃ MW: 209.2019 g/mol Melting Pt: 112…114 °C |
MDL Number:
MFCD00023308 CAS Number: 26820-62-2 |
Specification Test Results
Melting point | 112-114°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...