4-Acetyl-2-(trichloroacetyl)pyrrole ≥95%
Supplier: Matrix Scientific
Synonyms:
1-(4-Acetyl-1H-pyrrol-2-yl)-2,2,2-trichloroethan-1-one
045929-1G
045929-5G
045929-500MG
101873-312EA
556.92
USD
101873-312
101873-314
101873-316
4-Acetyl-2-(trichloroacetyl)pyrrole ≥95%
4-Acetyl-2-(trichloroacetyl)pyrrole
Hazard: Irritant
Formula:
C₈H₆Cl₃NO₂ Melting Pt: 201…203 °C |
MDL Number:
MFCD03001304 CAS Number: 72652-34-7 |
Specification Test Results
Melting point | 201-203°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...