2-Hydroxy-1,3,5-benzenetricarbaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Hydroxybenzene-1,3,5-tricarbaldehyde

045924-1G 045924-500MG 045924-5G
101873-284EA 195.38 USD
101873-284 101873-286 101873-288
2-Hydroxy-1,3,5-benzenetricarbaldehyde ≥95%
2-Hydroxy-1,3,5-benzenetricarbaldehyde

Hazard: Irritant

Formula: C₉H₆O₄
MW: 178.15 g/mol
Melting Pt: 201…202 °C
MDL Number: MFCD01314207
CAS Number: 81502-74-1

Order Now

Specification Test Results

Melting point 201-202°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR