4-(3-Formyl-2,5-dimethyl-1H-pyrrol-1-yl)benzenecarboxylic acid ≥95%

Supplier: Matrix Scientific

045907-500MG 045907-1G
101873-198EA 416.3 USD
101873-198 101873-200
4-(3-Formyl-2,5-dimethyl-1H-pyrrol-1-yl)benzenecarboxylic acid ≥95%
4-(3-Formyl-2,5-dimethyl-1H-pyrrol-1-yl)benzenecarboxylic acid

Hazard: Irritant

Formula: C₁₄H₁₃NO₃
Melting Pt: 259…261 °C
MDL Number: MFCD00022463
CAS Number: 52034-38-5

Order Now

Specification Test Results

Melting point 259-261°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR