4-Iodoisatin ≥95%
Supplier: Matrix Scientific
Synonyms:
4-Iodoindoline-2,3-dione
045906-1G
045906-500MG
101873-194EA
375.38
USD
101873-194
101873-196
4-Iodoisatin ≥95%
4-Iodoisatin
Hazard: Irritant
Formula:
C₈H₄INO₂ MW: 273.0300 g/mol Melting Pt: 227…229 °C |
MDL Number:
MFCD03427300 CAS Number: 20780-75-0 |
Specification Test Results
Melting point | 227-229°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...