Methyl-4-[(3-fluorobenzyl)oxy]benzenecarboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl 4-[(3-fluorobenzyl)oxy]benzoate
045892-5G
045892-500MG
045892-1G
101873-136EA
375.38
USD
101873-136
101873-138
101873-140
Methyl-4-[(3-fluorobenzyl)oxy]benzenecarboxylate ≥95%
Methyl-4-[(3-fluorobenzyl)oxy]benzenecarboxylate
Hazard: Irritant
Formula:
C₁₅H₁₃FO₃ MW: 260.27 g/mol Melting Pt: 71…73 °C |
MDL Number:
MFCD01568714 CAS Number: 454473-78-0 |
Specification Test Results
Melting point | 71-73°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...