2-Amino-5-oxo-4-(2-thienyl)-5,6,7,8-tetrahydro-4H-chromene-3-carbonitrile ≥95%

Supplier: Matrix Scientific

045863-500MG 045863-1G 045863-5G
101872-994EA 2129.23 USD
101872-994 101872-992 101872-990
2-Amino-5-oxo-4-(2-thienyl)-5,6,7,8-tetrahydro-4H-chromene-3-carbonitrile ≥95%
2-Amino-5-oxo-4-(2-thienyl)-5,6,7,8-tetrahydro-4H-chromene-3-carbonitrile

Hazard: Irritant

Formula: C₁₄H₁₂N₂O₂S
Melting Pt: 184…186 °C
MDL Number: MFCD00096530
CAS Number: 312510-11-5

Order Now

Specification Test Results

Melting point 184-186°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR