2-(Trifluoromethyl)-4(3H)-quinazolinone ≥95%

Supplier: Matrix Scientific

045858-5G 045858-500MG 045858-1G
101872-964EA 522.1 USD
101872-964 101872-962 101872-966
2-(Trifluoromethyl)-4(3H)-quinazolinone ≥95%
2-(Trifluoromethyl)-4(3H)-quinazolinone

Hazard: Irritant

Formula: C₉H₅F₃N₂O
Melting Pt: 248…251 °C
MDL Number: MFCD00793796
CAS Number: 26059-81-4

Order Now

Specification Test Results

Melting point 248-251°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR