1-(2,4-Dichlorobenzyl)-2-oxo-1,2-dihydro-3-pyridine carboxylic acid ≥95%
Supplier: Matrix Scientific
045816-1G
045816-5G
045816-500MG
101872-764EA
1384.62
USD
101872-764
101872-762
101872-760
1-(2,4-Dichlorobenzyl)-2-oxo-1,2-dihydro-3-pyridine carboxylic acid ≥95%
1-(2,4-Dichlorobenzyl)-2-oxo-1,2-dihydro-3-pyridine carboxylic acid
Hazard: Irritant
Formula:
C₁₃H₉Cl₂NO₃ MW: 298.13 g/mol Melting Pt: 177…178 °C |
MDL Number:
MFCD00140302 CAS Number: 338977-51-8 |
Specification Test Results
Melting point | 177-178°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...