4-(2,4-Dichlorobenzoyl)-N-(2-hydroxyethyl)-1H-pyrrole-2-carboxamide ≥95%

Supplier: Matrix Scientific

045815-1G 045815-5G 045815-500MG
101872-754EA 747.69 USD
101872-754 101872-758 101872-756
4-(2,4-Dichlorobenzoyl)-N-(2-hydroxyethyl)-1H-pyrrole-2-carboxamide ≥95%
4-(2,4-Dichlorobenzoyl)-N-(2-hydroxyethyl)-1H-pyrrole-2-carboxamide

Hazard: Irritant

Formula: C₁₄H₁₂Cl₂N₂O₃
Melting Pt: 160…162 °C
MDL Number: MFCD00141924
CAS Number: 338977-08-5

Order Now

Specification Test Results

Melting point 160-162°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR