4-(4-Fluorophenyl)-2-oxo-2,5-dihydro-3-furancarbonitrile ≥95%

Supplier: Matrix Scientific

045813-5G 045813-500MG 045813-1G
101872-744EA 153.85 USD
101872-744 101872-742 101872-746
4-(4-Fluorophenyl)-2-oxo-2,5-dihydro-3-furancarbonitrile ≥95%
4-(4-Fluorophenyl)-2-oxo-2,5-dihydro-3-furancarbonitrile

Hazard: Irritant

Formula: C₁₁H₆FNO₂
MW: 203.17 g/mol
Melting Pt: 210…212 °C
MDL Number: MFCD00139244
CAS Number: 50691-04-8

Order Now

Specification Test Results

Melting point 210-212°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR