4-[1,1'-Biphenyl]-4-yl-2-oxo-2,5-dihydro-3-furancarbonitrile ≥95%

Supplier: Matrix Scientific

045810-500MG 045810-5G 045810-1G
101872-726EA 467.69 USD
101872-726 101872-724 101872-728
4-[1,1'-Biphenyl]-4-yl-2-oxo-2,5-dihydro-3-furancarbonitrile ≥95%
4-[1,1'-Biphenyl]-4-yl-2-oxo-2,5-dihydro-3-furancarbonitrile

Hazard: Irritant

Formula: C₁₇H₁₁NO₂
Melting Pt: 244…247 °C
MDL Number: MFCD00139242
CAS Number: 50691-09-3

Order Now

Specification Test Results

Melting point 244-247°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR