2-Cyano-N-[3-(trifluoromethyl)phenyl]acetamide ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Cyano-3'-(trifluoromethyl)acetanilide

045798-500MG 045798-1G 045798-5G
101872-656EA 232.31 USD
101872-656 101872-654 101872-658
2-Cyano-N-[3-(trifluoromethyl)phenyl]acetamide ≥95%
2-Cyano-N-[3-(trifluoromethyl)phenyl]acetamide

Hazard: Irritant

Formula: C₁₀H₇F₃N₂O
MW: 228.18 g/mol
Melting Pt: 143…145 °C
MDL Number: MFCD00117941
CAS Number: 1960-77-6

Order Now

Specification Test Results

Melting point 143-145°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR