8-Nitroimidazo[1,2-a]pyridine ≥97%

Supplier: Matrix Scientific

045790-5G 045790-1G 045790-500MG
101872-612EA 140 USD
101872-612 101872-616 101872-614
8-Nitroimidazo[1,2-a]pyridine ≥97%
8-Nitroimidazo[1,2-a]pyridine

Hazard: Irritant

Formula: C₇H₅N₃O₂
MW: 163.1335 g/mol
Melting Pt: 188…190 °C
MDL Number: MFCD08056674
CAS Number: 52310-46-0

Order Now

Specification Test Results

Melting point 188-190°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR