2-(Methylsulfanyl)-1,4,5,6-tetrahydropyrimidinehydroiodide ≥95%
Supplier: Matrix Scientific
Synonyms:
2-(Methylthio)-1,4,5,6-tetrahydropyrimidine hydroiodide
045788-500MG
045788-1G
045788-5G
101872-604EA
369.23
USD
101872-604
101872-602
101872-606
2-(Methylsulfanyl)-1,4,5,6-tetrahydropyrimidinehydroiodide ≥95%
2-(Methylsulfanyl)-1,4,5,6-tetrahydropyrimidinehydroiodide
Hazard: Irritant
Formula:
C₅H₁₁IN₂S MW: 258.13 g/mol Melting Pt: 138…139 °C |
MDL Number:
MFCD00829842 CAS Number: 5445-73-8 |
Specification Test Results
Melting point | 138-139°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...