5-Nitro-2-piperazinobenzenecarboxylic acid ≥95%

Supplier: Matrix Scientific

045776-1G 045776-500MG
101872-534EA 698.46 USD
101872-534 101872-532
5-Nitro-2-piperazinobenzenecarboxylic acid ≥95%
5-Nitro-2-piperazinobenzenecarboxylic acid

Hazard: Irritant

Formula: C₁₁H₁₃N₃O₄
Melting Pt: 295 °C (dec.)
MDL Number: MFCD06659596
CAS Number: 168123-39-5

Order Now

Specification Test Results

Melting point 295°C(dec)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR