(2-Amino-3-thienyl)(phenyl)methanone ≥95%
Supplier: Matrix Scientific
045764-500MG
101872-472EA
278.46
USD
101872-472
(2-Amino-3-thienyl)(phenyl)methanone ≥95%
(2-Amino-3-thienyl)(phenyl)methanone
Hazard: Irritant
Formula:
C₁₁H₉NOS Melting Pt: 149…151 °C |
MDL Number:
MFCD01927388 CAS Number: 21582-44-5 |
Specification Test Results
Melting point | 149-151°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...