4-[(3-Fluorobenzyl)oxy]benzenecarbohydrazide ≥95%
Supplier: Matrix Scientific
Synonyms:
4-(3-Fluorobenzyloxy)benzhydrazide
045749-500MG
045749-5G
045749-1G
101872-414EA
1600
USD
101872-414
101872-412
101872-410
4-[(3-Fluorobenzyl)oxy]benzenecarbohydrazide ≥95%
4-[(3-Fluorobenzyl)oxy]benzenecarbohydrazide
Hazard: Irritant
Formula:
C₁₄H₁₃FN₂O₂ MW: 260.27 g/mol Melting Pt: 118…120 °C |
MDL Number:
MFCD05663827 CAS Number: 886361-23-5 |
Specification Test Results
Melting point | 118-120°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...