4-(4-Methylbenzoyl)-1H-pyrrole-2-carboxylic acid ≥95%
Supplier: Matrix Scientific
045744-500MG
045744-1G
101872-400EA
927.69
USD
101872-400
101872-398
4-(4-Methylbenzoyl)-1H-pyrrole-2-carboxylic acid ≥95%
4-(4-Methylbenzoyl)-1H-pyrrole-2-carboxylic acid
Hazard: Irritant
Formula:
C₁₃H₁₁NO₃ Melting Pt: 247…249 °C |
MDL Number:
MFCD05663824 CAS Number: 886361-19-9 |
Specification Test Results
Melting point | 247-249°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...