3-[2-(2,4-Dichlorophenoxy)phenyl]propanoic acid ≥95%

Supplier: Matrix Scientific

045738-1G 045738-5G 045738-500MG
101872-368EA 921.54 USD
101872-368 101872-372 101872-370
3-[2-(2,4-Dichlorophenoxy)phenyl]propanoic acid ≥95%
3-[2-(2,4-Dichlorophenoxy)phenyl]propanoic acid

Hazard: Irritant

Formula: C₁₅H₁₂Cl₂O₃
Melting Pt: 104…107 °C
MDL Number: MFCD05663818
CAS Number: 86308-89-6

Order Now

Specification Test Results

Melting point 104-107°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR