4,5-Dihydronaphtho[1,2-b]thiophene-2-carbohydrazide ≥95%
Supplier: Matrix Scientific
045735-500MG
045735-5G
045735-1G
101872-350EA
1109.23
USD
101872-350
101872-354
101872-352
4,5-Dihydronaphtho[1,2-b]thiophene-2-carbohydrazide ≥95%
4,5-Dihydronaphtho[1,2-b]thiophene-2-carbohydrazide
Hazard: Irritant
Formula:
C₁₃H₁₂N₂OS Melting Pt: 162…165 °C |
MDL Number:
MFCD01764165 CAS Number: 278782-13-1 |
Specification Test Results
Melting point | 162-165°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...