5-Bromo-1-(2,4-dichlorobenzyl)-2-oxo-1,2-dihydro-3-pyridine carboxylic acid ≥95%
Supplier: Matrix Scientific
045726-500MG
045726-1G
045726-5G
101872-318EA
698.46
USD
101872-318
101872-316
101872-320
5-Bromo-1-(2,4-dichlorobenzyl)-2-oxo-1,2-dihydro-3-pyridine carboxylic acid ≥95%
5-Bromo-1-(2,4-dichlorobenzyl)-2-oxo-1,2-dihydro-3-pyridine carboxylic acid
Hazard: Irritant
Formula:
C₁₃H₈BrCl₂NO₃ Melting Pt: 220…223 °C |
MDL Number:
MFCD05663811 CAS Number: 886360-99-2 |
Specification Test Results
Melting point | 220-223°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...